CAS RN 135159-51-2
IUPAC Name:
hydrogen 4-{[1-(dimethylamino)-3-{2-[2-(3-methoxyphenyl)ethyl]phenoxy}propan-2-yl]oxy}-4-oxobutanoic acid chloride
Synonyms:
hydrogen sarpogrelate chloride
Molecular Weight (g/mol):
465.97
Molecular Formula:
C24H32ClNO6
InChi Key:
POQBIDFFYCYHOB-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC1=CC=CC(CCC2=CC=CC=C2OCC(CN(C)C)OC(=O)CCC(O)=O)=C1
1
–
1
of
1
results
1
Thermo Scientific™ Sarpogrelate hydrochloride, 98%
CAS: 135159-51-2 Molecular Formula: C24H31NO6·HCl Molecular Weight (g/mol): 465.97 InChI Key: POQBIDFFYCYHOB-UHFFFAOYNA-N PubChem CID: 444005