missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sarpogrelate hydrochloride, 98%

Catalog No. AC461912500
Change view
Click to view available options
Quantity:
250mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC461912500 250mg
AC461910010 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC461912500 Supplier Thermo Scientific™ Supplier No. 461912500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 135159-51-2
Molecular Formula C24H31NO6·HCl
Molecular Weight (g/mol) 465.97
InChI Key POQBIDFFYCYHOB-UHFFFAOYNA-N
PubChem CID 444005

Specifications

Assay Percent Range 97.5% min. (HPLC)
Quantity 250mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dimethylsulfoxide and methanol
Formula Weight 465.97
Percent Purity 98%
Chemical Name or Material Sarpogrelate hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Very toxic to aquatic life.

RUO – Research Use Only