missing translation for 'onlineSavingsMsg'
Learn More

Sarcosine Ethyl Ester Hydrochloride 98.0+%, TCI America™

Catalog No. p-7135958
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S08415G 5 g
S084125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S08415G Supplier TCI America Supplier No. S08415G

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
IUPAC Name hydrogen ethyl 2-(methylamino)acetate chloride
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 125°C
Color White
Quantity 5 g
Formula Weight 153.61
Percent Purity ≥98.0% (N,T)
Physical Form Crystalline Powder
Chemical Name or Material Sarcosine Ethyl Ester Hydrochloride