missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Sarcosine ethyl ester hydrochloride, 97%

Catalog No. AC305590100
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC305590100 10 g Glass bottle
AC305590500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC305590100 Supplier Thermo Scientific Chemicals Supplier No. 305590100
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 52605-49-9 | C5H12ClNO2 | 153.61 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 52605-49-9
Molecular Formula C5H12ClNO2
Molecular Weight (g/mol) 153.61
MDL Number MFCD00012506
InChI Key NIDZUMSLERGAON-UHFFFAOYSA-N
Synonym sarcosine ethyl ester hydrochloride, ethyl sarcosinate hydrochloride, ethyl 2-methylamino acetate hydrochloride, h-sar-oet.hcl, ethyl n-methylaminoacetate hydrochloride, me-gly-oet.hcl, n-methylglycine ethyl ester hydrochloride, glycine, n-methyl-, ethyl ester, hydrochloride, ethyl n-methylglycinate hydrochloride, n-methyl glycine ethyl ester hydrochloride
PubChem CID 171173
SMILES [H+].[Cl-].CCOC(=O)CNC

Specifications

Melting Point 123°C to 129°C
Color White
Loss on Drying 1% max.
Infrared Spectrum Authentic
Assay Percent Range 96% min. (Argentometry)
Packaging Glass bottle
Linear Formula CH3NHCH2CO2C2H5·HCl
Quantity 10 g
IUPAC Name ethyl 2-(methylamino)acetate;hydrochloride
Formula Weight 153.61
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material Sarcosine ethyl ester hydrochloride
Show More Show Less
EINECSNumber 258-037-5

RUO – Research Use Only