missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sarafloxacin Hydrochloride 97.0+%, TCI America™
$154.71 - $154.71
Chemical Identifiers
| CAS | 91296-87-6 |
|---|---|
| Molecular Formula | C20H18ClF2N3O3 |
| Molecular Weight (g/mol) | 421.829 |
| MDL Number | MFCD11869789 |
| InChI Key | KNWODGJQLCISLC-UHFFFAOYSA-N |
| Synonym | sarafloxacin hydrochloride, sarafloxacin hcl, unii-i36jp4q9df, abbott-56620, i36jp4q9df, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237 |
| PubChem CID | 56207 |
| IUPAC Name | 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride |
| SMILES | C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl |
Chemical Identifiers
| 91296-87-6 | |
| 421.829 | |
| KNWODGJQLCISLC-UHFFFAOYSA-N | |
| 56207 | |
| C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl |
| C20H18ClF2N3O3 | |
| MFCD11869789 | |
| sarafloxacin hydrochloride, sarafloxacin hcl, unii-i36jp4q9df, abbott-56620, i36jp4q9df, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237 | |
| 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride |
Specifications
| 91296-87-6 | |
| MFCD11869789 | |
| sarafloxacin hydrochloride, sarafloxacin hcl, unii-i36jp4q9df, abbott-56620, i36jp4q9df, sarafloxacin hydrochloride usan, saraflox, dsstox_cid_25939, dsstox_rid_81237 | |
| C1CN(CCN1)C2=C(C=C3C(=C2)N(C=C(C3=O)C(=O)O)C4=CC=C(C=C4)F)F.Cl | |
| 421.829 | |
| 421.83 | |
| Crystalline Powder |
| C20H18ClF2N3O3 | |
| 1 g | |
| KNWODGJQLCISLC-UHFFFAOYSA-N | |
| 6-fluoro-1-(4-fluorophenyl)-4-oxo-7-piperazin-1-ylquinoline-3-carboxylic acid;hydrochloride | |
| 56207 | |
| ≥97.0% (HPLC) | |
| Sarafloxacin Hydrochloride |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator