missing translation for 'onlineSavingsMsg'
Learn More

SAPS2 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501558913
Encompass
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501558913 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 501558913 Supplier Bethyl Laboratories Inc Supplier No. A300969A

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

Regulatory subunit of protein phosphatase 6 (PP6). May function as a scaffolding PP6 subunit. Involved in the PP6- mediated dephosphorylation of NFKBIE opposing its degradation in response to TNF-alpha.

Specifications

Antigen SAPS2
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.2 mg/ml
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene PPP6R2
Gene Accession No. O75170
Gene Alias Serine/threonine-protein phosphatase 6 regulatory subunit 2, SAPS domain family member 2, PPP6R2, KIAA0685, PP6R2, SAPS2
Gene Symbols PPP6R2
Host Species Rabbit
Immunogen Between 775 and 825
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9701
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less