Learn More
Azure Biosystems Sapphire FL Standard Optical Module
Catalog No. IS4001AZ
Change view
Click to view available options
Wavelength:
488 nm
532 nm
638 nm
658 nm
685 nm
784 nm
Includes:
513/17 nm Filter
534/20 nm Filter
572/28 nm Filter
624/40 nm Filter
676/37 nm Filter
710/40 nm Filter
720/24 nm Filter
829/62 nm Filter
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Includes
IS4001AZ 488 nm 513/17 nm Filter
IS4002AZ 532 nm 572/28 nm Filter
IS4057AZ 658 nm 710/40 nm Filter
IS4003AZ 638 nm 676/37 nm Filter
IS4004AZ 685 nm 720/24 nm Filter
IS4005AZ 784 nm 829/62 nm Filter
IS4032AZ 488 nm 534/20 nm Filter
IS4033AZ 532 nm 624/40 nm Filter
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. IS4001AZ Supplier Azure Biosystems Supplier No. IS4001
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Includes laser and emission filter for use with Sapphire FL
  • Standard Optical Module for Sapphire FL
  • Includes laser and emission filter.
Specifications
Includes 513/17 nm Filter
Wavelength 488 nm
Product Type Optical Module