Learn More
Azure Biosystems Sapphire FL Slide Holder
Description
- The holder is designed to hold up to 24 standard microscope slides at once. This significantly increases throughput for slide-based assays
- It is an accessory for the Azure Biosystems Sapphire FL and Sapphire FL+ Biomolecular Imagers
- Adjustable Z-Plane Focus, available in black color
Specifications
Specifications
| Type | Slide Holder |
| Slide Capacity | 24 |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Sterility | Non-sterile |