missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azure Biosystems Sapphire FL Chemiluminescent Module Ethernet Connection Kit
Description
- Contains 7 Foot Cat6 (UTP) Ethernet Cable and 4 port Netgear ethernet switch.
Specifications
Specifications
| For Use With (Equipment) | Sapphire FL |
| Product Type | Ethernet Connection |