missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAPAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520541
Description
A suggested positive control is L1210 cell lysate. PA5-20541 can be used with blocking peptide PEP-0661.
SAP90/PSD-95-associated protein 2 (SAPAP2, also known as DLGAP2) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP2 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP2 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -4, whereas SAPAP3 mRNA is detected mainly in cell bodies.Specifications
| SAPAP2 | |
| Polyclonal | |
| Unconjugated | |
| DLGAP2 | |
| 6430596N04Rik; Dap2; DAP-2; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; discs, large (Drosophila) homolog-associated protein 2; disks large-associated protein 2; DLG associated protein 2; Dlgap2; postsynaptic density 95; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 2; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 2; SAPAP2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116681, 244310, 9228 | |
| Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| P97837, Q8BJ42, Q9P1A6 | |
| DLGAP2 | |
| A 16 amino acid peptide from near the center of human SAPAP2. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |