missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAPAP2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA556438
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA556438 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA556438 Supplier Invitrogen™ Supplier No. PA556438
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Rabbit Polyclonal Antibody

Immunogen sequence: HSKRSKSKER KPEGKPRPGM SSWWSSDDNL DSDSTYRTPS VLNRHHLGPV AHCYPDALQS PFGDLSLKTS KSNNDVKCSA CEGLALTPDA KYLKRSSWST Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 84%.

SAP90/PSD-95-associated protein 2 (SAPAP2, also known as DLGAP2) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP2 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP2 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -4, whereas SAPAP3 mRNA is detected mainly in cell bodies.

Specifications

Antigen SAPAP2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene DLGAP2
Gene Accession No. Q9P1A6
Gene Alias 6430596N04Rik; Dap2; DAP-2; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; discs, large (Drosophila) homolog-associated protein 2; disks large-associated protein 2; DLG associated protein 2; Dlgap2; postsynaptic density 95; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 2; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 2; SAPAP2
Gene Symbols DLGAP2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAPAP2. Recombinant protein control fragment (Product #RP-93954).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9228
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less