missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAPAP2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA532172
Description
Recommended positive controls: Mouse brain. Predicted reactivity: Mouse (100%), Rat (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
SAP90/PSD-95-associated protein 2 (SAPAP2, also known as DLGAP2) is a member of a protein family whose members specifically interact with PSD-95/SAP90, a membrane-associated guanylate kinase localized at postsynaptic density (PSD) in neuronal cells. Like the other SAPAP proteins, SAPAP2 is thought to be an adaptor protein that also interacts with different synaptic scaffolding proteins, cytoskeletal and signaling components, such as focal adhesion kinase (FAK) and proline-rich tyrosine kinase 2 (PYK2). SAPAP2 mRNA is targeted to cell bodies in a similar manner to SAPAP1 and -4, whereas SAPAP3 mRNA is detected mainly in cell bodies.Specifications
| SAPAP2 | |
| Polyclonal | |
| Unconjugated | |
| DLGAP2 | |
| 6430596N04Rik; Dap2; DAP-2; discs large homolog associated protein 2; discs large homolog-associated protein 2; discs large-associated protein 2; discs, large (Drosophila) homolog-associated protein 2; disks large-associated protein 2; DLG associated protein 2; Dlgap2; postsynaptic density 95; PSD-95/SAP90-associated protein-2; PSD-95/SAP90-binding protein 2; SAP90/PSD-95 associated protein 2; SAP90/PSD-95-associated protein 2; SAPAP2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 244310 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8BJ42 | |
| DLGAP2 | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the N-terminus region of mouse Dlgap2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |