missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAP130 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA589514
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA589514 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA589514 Supplier Invitrogen™ Supplier No. PA589514
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: LSMPTSDLPP GASPRKKPRK QQHVISTEEG DMMETNSTDD EKSTAKSLLV KAEKRKSPPK EYIDEEGVRY VPVRPRPPIT LLRHYRNPWK AAYHHFQRYS DVRVKEEKKA MLQEIANQKG VSCRAQGWKV HLCAAQLLQL TNLEHDVYER LTNLQEGIIP KKKAATDDDL HRINELIQGN MQRCKLVMDQ ISEARDSMLK VLDHKDRVLK LLNKNGTVKK VSKLKRKEKV; Positive Samples: Mouse brain; Cellular Location: Nucleus.

SAP130 is a subunit of the histone deacetylase-dependent SIN3A corepressor complex (Fleischer et al., 2003).

Specifications

Antigen SAP130
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.56 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene Sap130
Gene Accession No. Q8BIH0
Gene Alias 130 kDa Sin3-associated polypeptide; 2610304F09Rik; 6720406D06; FLJ12761; Histone deacetylase complex subunit SAP130; KIAA0017; mSin3A-associated protein 130; pre-mRNA splicing factor SF3b, 130 kDa subunit; pre-mRNA-splicing factor SF3b 130 kDa subunit; RGD1311657; RSE1; SAP 130; SAP130; SF3b130; SF3B3; Sin3A associated protein; Sin3A associated protein 130; Sin3A associated protein 130kDa; Sin3A associated protein, 130kDa; Sin3A-associated protein, 130kDa; Sin3-associated polypeptide p130; spliceosome-associated protein 130; Splicing factor 3B subunit 3; STAF130
Gene Symbols Sap130
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 819-1048 of human SAP130 (NP_0788212).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 269003
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less