missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Ottawa, Standard, 20-30 Mesh, Spectrum™ Chemical
S1010, 14808-60-7
$211.43 - $2001.03
Chemical Identifiers
| CAS | 14808-60-7 |
|---|---|
| Molecular Formula | O2Si |
| Molecular Weight (g/mol) | 60.08 |
| MDL Number | MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 |
| InChI Key | VYPSYNLAJGMNEJ-UHFFFAOYSA-N |
| IUPAC Name | silanedione |
| SMILES | O=[Si]=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18606164
|
Spectrum Chemical Mfg Cor
S1010500GM |
500 g | Poly Bottle |
Each for $211.43
|
|
||||
|
18606165
|
Spectrum Chemical Mfg Cor
S101025KG |
2.5 kg | Poly Bottle |
Each for $611.37
|
|
||||
|
18606166
|
Spectrum Chemical Mfg Cor
S101045KGBL |
45 kg | Fiber Drum |
Each for $2,001.03
|
|
||||
Description
Spectrum™ Chemical Sand, Ottawa, Standard, 20-30 MeshChemical Identifiers
| 14808-60-7 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| silanedione |
Specifications
| 14808-60-7 | |
| 500 g | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| O=[Si]=O | |
| 60.08 | |
| Poly Bottle |
| 1 | |
| O2Si | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| Reagent |