missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, Ottawa, Standard, 20-30 Mesh, Spectrum™ Chemical
S1010, 14808-60-7
Supplier: Spectrum Chemical Mfg Cor S101025KG
Description
Spectrum™ Chemical Sand, Ottawa, Standard, 20-30 MeshSpecifications
| 14808-60-7 | |
| 2.5 kg | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| O=[Si]=O | |
| 60.08 | |
| Poly Bottle |
| 1 | |
| O2Si | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| silanedione | |
| Reagent |
Chemical Identifiers
| 14808-60-7 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| O=[Si]=O |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| silanedione |