missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sand, for analysis, 40-100 mesh
CAS: 14808-60-7 | O2Si | 60.08 g/mol
Supplier: Thermo Scientific Chemicals 393300010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Filter aidSpecifications
| 2230.0°C | |
| 1610.0°C | |
| 14808-60-7 | |
| 1 kg | |
| SiO2 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| dioxosilane | |
| 24261 | |
| 60.08 | |
| Plastic bottle |
| Sand | |
| Beige to Gray | |
| Crystals | |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| Solubility in water: insoluble | |
| O=[Si]=O | |
| 60.08 | |
| CHEBI:30563 | |
| Analytical |
Chemical Identifiers
| 14808-60-7 | |
| 60.08 | |
| VYPSYNLAJGMNEJ-UHFFFAOYSA-N | |
| 24261 | |
| dioxosilane |
| O2Si | |
| MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733 | |
| silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous | |
| CHEBI:30563 | |
| O=[Si]=O |
Safety and Handling
EINECSNumber : 238-878-4
RUO – Research Use Only