missing translation for 'onlineSavingsMsg'
Learn More

Sand, for analysis, 40-100 mesh

Catalog No. p-3586493
Change view
Click to view available options
Quantity:
250 g
1 kg
5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC393302500 250 g Plastic bottle
AC393300010 1 kg Plastic bottle
AC393300050 5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC393302500 Supplier Thermo Scientific Chemicals Supplier No. 393302500
Only null left

CAS: 14808-60-7 | O2Si | 60.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Filter aid

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
MDL Number MFCD00011232,MFCD00217788,MFCD00163736,MFCD07370733
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
Synonym silica, silicon dioxide, quartz, cristobalite, diatomaceous earth, tridymite, silicic anhydride, aerosil, infusorial earth, silica, amorphous
PubChem CID 24261
ChEBI CHEBI:30563
IUPAC Name dioxosilane
SMILES O=[Si]=O

Specifications

Boiling Point 2230.0°C
Melting Point 1610.0°C
Color Beige to Gray
Physical Form Crystals
Quantity 250 g
Linear Formula SiO2
Solubility Information Solubility in water: insoluble
Formula Weight 60.08
Grade Analytical
Packaging Plastic bottle
Chemical Name or Material Sand
Show More Show Less
EINECSNumber 238-878-4

RUO – Research Use Only