Learn More
Globe Scientific Sample Cup for Elan Diagnostics Series Analyzers
Designed for use on the Elan Atac 6000 & 8000 analyzers. Globe Scientific Sample Cups for Elan Diagnostics Series Analyzers are constructed from polypropylene and are designed for compatibility with these products.
Description
- For use on the Elan Atac 6000 & 8000 analyzers
- Designed for compatibility with these products
- Polypropylene (PP) construction
- White color
Specifications
| White | |
| 1000 | |
| Sample Cup |
| Polypropylene | |
| Elan Atac 6000 and 8000 Analyzers |