missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAMHD1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583852
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583852 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583852 Supplier Invitrogen™ Supplier No. PA583852
Only null left

Rabbit Polyclonal Antibody

Immunogen sequence: DDSPRTPSNT PSAEADWSPG LELHPDYKTW GPEQVCSFLR RGGFEEPVLL KNIRENEITG ALLPCLDESR FENLGVSSLG ERKKLLSYIQ RLVQIHVDTM KVI Highest antigen sequence indentity to the following orthologs: Mouse - 52%, Rat - 52%.

This gene may play a role in regulation of the innate immune response. The encoded protein is upregulated in response to viral infection and may be involved in mediation of tumor necrosis factor-alpha proinflammatory responses. Mutations in this gene have been associated with Aicardi-Goutieres syndrome.

Specifications

Antigen SAMHD1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SAMHD1
Gene Accession No. Q9Y3Z3
Gene Alias CHBL2; DCIP; Dendritic cell-derived IFNG-induced protein; deoxynucleoside triphosphate triphosphohydrolase SAMHD1; dNTPase; E330031J07Rik; EGK_02438; HDDC1; hSAMHD1; IFN-gamma induced; interferon-gamma-inducible protein Mg11; Mg11; Mg21; Monocyte protein 5; MOP5; MOP-5; mSAMHD1; RP1-132F21.1; SAM and HD domain containing deoxynucleoside triphosphate triphosphohydrolase 1; SAM domain and HD domain 1; SAM domain and HD domain, 1; SAM domain and HD domain-containing protein 1; Samhd1; SBBI88
Gene Symbols SAMHD1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SAMHD1. Recombinant protein control fragment (Product #RP-99320).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25939
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less