missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SAMD12 Polyclonal Antibody
SAMD12 Polyclonal Antibody
Supplier: Thermo Fisher Scientific PA555131
Description
SAMD12 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| SAMD12 | |
| Polyclonal | |
| Unconjugated | |
| SAMD12 | |
| SAM domain-containing protein 12; SAMD12; sterile alpha motif domain containing 12; sterile alpha motif domain-containing protein 12 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 401474 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N8I0 | |
| SAMD12 | |
| Recombinant protein corresponding to Human SAMD12. Recombinant protein control fragment (Product #RP-93966). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |