missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SAMD12 Monoclonal Antibody (OTI2G1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527084
Description
SAMD12 Monoclonal Antibody for Western Blot, IHC (P)
SAMD12 is a protein coding gene.Specifications
| SAMD12 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8N8I0 | |
| SAMD12 | |
| Full length human recombinant protein of SAMD12 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G1 | |
| Unconjugated | |
| SAMD12 | |
| SAM domain-containing protein 12; SAMD12; sterile alpha motif domain containing 12; sterile alpha motif domain-containing protein 12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 401474 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |