missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers

Prevent bio-hazardous spills while conserving laboratory space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.

$439.75 - $692.97

Spécifications

Product Type Specimen Container
Closure Included Yes
Material Virgin Polypropylene
Capacity (Metric) 90 mL
Closure Size 48 mm
voir les spécifications

Produits 5
Numéro de catalogue Numéro du manufacturier. Sterility Label Closure Color No. per Case Prix Quantité  
Numéro de catalogue Numéro du manufacturier. Sterility Label Closure Color No. per Case Prix Quantité  
13-711-61
Afficher les documents
Thermo Scientific™
020016
Sterile Tabbed Orange 400/Cs. (bulk)
caisse de 400 chaque for $541.33
Il en reste null
 
13-711-74
Afficher les documents
Thermo Scientific™
020214
Sterile Tabbed Orange 400/Cs.
caisse de 400 chaque for $692.97
Il en reste null
 
22-045365
Afficher les documents
Thermo Scientific™
020038
Sterile Tabbed Natural 400/Cs.
caisse de 400 chaque for $520.22
Il en reste null
 
13-711-58
Afficher les documents
Thermo Scientific™
020039
Sterile Tabbed White 400/Cs.
caisse de 400 chaque for $538.81
Il en reste null
 
13-711-71
Afficher les documents
Thermo Scientific™
020148
Non-sterile Non-tabbed Yellow 400/Cs.
caisse de 400 chaque for $439.75
Il en reste null
 
Description

Description

Prevent bio-hazardous spills while conserving laboratory space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.

Features:
  • Dual-threaded caps create leak-resistant seal for superior safety and handling
  • Top and bottom positive grip knurls are easy to grip and open using latex gloves
  • Virgin polypropylene allows higher clarity to see samples, and will not become brittle
  • Sterile indicator tab assures tamper evidence
  • Bilingual English and Spanish label facilitates accurate usage

Ordering Information:

  • Available nonsterile or gamma irradiation sterilized
  • Available labeled or non-labeled
  • Available bulk or individually packed
  • In cases of 400
Spécifications

Spécifications

Specimen Container
Virgin Polypropylene
48 mm
48 mm
Yes
90 mL
Bio-Tite Lid
Videos
FDS
missing translation for 'documents'

missing translation for 'documents'