Learn More
Thermo Scientific™ Samco™ Narrow Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers
Prevent bio-hazardous spills while conserving laboratory space with these stackable containers, featuring a proprietary, all plastic, leak-resistant seal.
Supplier: Thermo Scientific™ 020016
Description
Prevent bio-hazardous spills while conserving laboratory space with stackable Thermo Scientific™ Samco™ Narrow-Mouth Bio-Tite™ 90mL (3 oz.) 48mm Specimen Containers. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
- Sterile indicator tab assures tamper evidence
- Bilingual English and Spanish label facilitates accurate usage
Ordering Information:
- Available nonsterile or gamma irradiation sterilized
- Available labeled or non-labeled
- Available bulk or individually packed
- In cases of 400
Specifications
| Specimen Container | |
| Virgin Polypropylene | |
| Sterile | |
| Orange | |
| Bio-Tite Lid | |
| 48 mm | |
| 400/Cs. (bulk) |
| Yes | |
| Cylindrical | |
| 90 mL | |
| 48 mm | |
| Sterile; Tabbed label; Orange caps; Bulk | |
| Tabbed |