Learn More
Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers

Prevent bio-hazardous spills and conserve storage space with these stackable containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings.
$368.06 - $469.06
Specifications
| Product Type | Specimen Container |
|---|---|
| Material | Virgin Polypropylene |
| Shape | Cylindrical |
| Sterility | Non-sterile |
| Closure Type | Bio-Tite Lid |
Description
Prevent bio-hazardous spills and conserve storage space with stackable Thermo Scientific™ Samco™ Histology Bio-Tite™ Nonsterile Specimen Containers, offered in capacities from 20mL to 120mL with 35, 48 or 53mm openings. These shatter-resistant containers feature a proprietary, all plastic, leak-resistant seal. They are tested at varying pressures, making them ideal for use in various transport systems.
Features:- Dual-threaded caps create leak-resistant seal for superior safety and handling
- Top and bottom positive grip knurls are easy to grip and open using latex gloves
- Virgin polypropylene allows higher clarity to see samples, and will not become brittle
Ordering Information:
Bulk packed, one bag per case. Caps must be ordered separately.
Specifications
| Specimen Container | |
| Cylindrical | |
| Bio-Tite Lid |
| Virgin Polypropylene | |
| Non-sterile | |
| No Label |