missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Samarium Standard For ICP, 1000μg/mL in 2% HNO3, Assurance Grade, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
$275.59 - $500.50
Chemical Identifiers
| CAS | 7440-19-9 |
|---|---|
| Molecular Formula | Sm |
| Molecular Weight (g/mol) | 150.36 |
| MDL Number | MFCD00011233 MFCD00151299 |
| InChI Key | KZUNJOHGWZRPMI-UHFFFAOYSA-N |
| PubChem CID | 23951 |
| ChEBI | CHEBI:33374 |
| IUPAC Name | samarium |
| SMILES | [Sm] |
Includes: NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
PLSM2-2Y
|
SPEX CertiPrep Group LLC
PLSM22Y |
125 mL | Triple-leached LDPE bottle |
Each for $275.59
|
|
||||
|
PLSM2-2X
|
SPEX CertiPrep Group LLC
PLSM22X |
500 mL | Triple-leached LDPE bottle |
Each for $500.50
|
|
||||
Description
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Sm | 1000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Chemical Identifiers
| 7440-19-9 | |
| 150.36 | |
| KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
| CHEBI:33374 | |
| [Sm] |
| Sm | |
| MFCD00011233 MFCD00151299 | |
| 23951 | |
| samarium |
Specifications
| Triple-leached LDPE bottle | |
| Assurance Certificate of Analysis reports up to 39 elements at ppb levels. | |
| 0.1,2.0,97.9 | |
| Sm | |
| AA, ICP | |
| Certified Reference Material | |
| [Sm] | |
| samarium | |
| 125 mL | |
| CHEBI:33374 | |
| Matrix Concentration Cat. No. 2-5% HNO3 1000μg/mL PLSM2-2Y, -2X 10,000 PLSM2-3Y, -3X |
| 7440-19-9,7697-37-2,7732-18-5 | |
| Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis. | |
| 0.1,2.0,97.9 | |
| MFCD00011233 MFCD00151299 | |
| AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation | |
| KZUNJOHGWZRPMI-UHFFFAOYSA-N | |
| 2% HNO3 | |
| 150.36 | |
| 23951 | |
| Assurance | |
| Samarium Standard |
Safety and Handling
Toxic in contact with skin. Causes severe skin burns and eye damage
DOTInformation : Hazardous
Recommended Storage : Ambient
Product intended for laboratory use only