Learn More
Samarium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific™
$80.85 - $241.85
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 14913-18-9 | |
| MFCD05665148 |
Specifications
| 14913-18-9 | |
| 2 g | |
| 246.41 (Anhydrous) | |
| Samarium(III) phosphate hydrate |
| Powder | |
| MFCD05665148 | |
| 99.9% | |
| (REO) |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 236-698-0
TSCA : No
Recommended Storage : Ambient temperatures