CAS RN 14913-18-9
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Samarium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific™
CAS: 14913-18-9 MDL Number: MFCD05665148