missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) phosphate hydrate, 99.9% (REO), Thermo Scientific™

Catalog No. AA4199209
Change view
Click to view available options
Quantity:
2 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4199209 10 g
AA4199204 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4199209 Supplier Thermo Scientific Chemicals Supplier No. 4199209

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 14913-18-9
MDL Number MFCD05665148

Specifications

Physical Form Powder
Quantity 10 g
Formula Weight 246.41 (Anhydrous)
Percent Purity 99.9%
Assay (REO)
Chemical Name or Material Samarium(III) phosphate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 236-698-0
TSCA No
Recommended Storage Ambient temperatures