missing translation for 'onlineSavingsMsg'
Learn More

Samarium(III) oxide, REacton™, 99.9% (REO)

Catalog No. p-4408157
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1122914 25 g
AA1122922 100 g
AA1122936 500 g
AA11229A1 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA1122914 Supplier Thermo Scientific Chemicals Supplier No. 01122914
Only null left

CAS: 12060-58-1 | Sm2O3

Samarium(III) oxide is used in optical and infrared absorbing glass to absorb infrared radiation. It finds use as a neutron absorber in nuclear power reactors to control the reaction. It is also used as a catalyst for the dehydration of primary alcohols to aldehydes or ketones. It forms samarium perchlorate when treated with perchloric acid, which is used as a reference material for checking the wavelength scale of a spectrophotometer in the range of 200 to 500 nm.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12060-58-1
Molecular Formula Sm2O3
MDL Number MFCD00011237

Specifications

Melting Point 2300°C
Color White
Physical Form Powder
Quantity 25 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 348.7
Refractive Index 1.97
Appearance Off-white powder
Density 8.347g/cm3 at 20°C
Chemical Name or Material Samarium(III) oxide
Show More Show Less
EINECSNumber 235-043-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only