missing translation for 'onlineSavingsMsg'
Learn More

Samarium cobalt, REacton™, Sm 33%

Catalog No. AA4273230
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4273230 250 g
AA4273218 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA4273230 Supplier Thermo Scientific Chemicals Supplier No. 04273230

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12017-68-4 | CoSm | 209.293 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12017-68-4
Molecular Formula CoSm
Molecular Weight (g/mol) 209.293
MDL Number MFCD02091682
InChI Key KPLQYGBQNPPQGA-UHFFFAOYSA-N
Synonym samarium cobalt, samarium cobalt powder,-30 mesh, cobalt-samarium, samarium-cobalt, powder30mesh, smco5, acmc-20ajrh, cobalt samarium, samarium cobalt, powder, sm2co17
PubChem CID 10125161
IUPAC Name cobalt;samarium
SMILES [Co].[Sm]

Specifications

Physical Form Fine Powder
Quantity 250 g
UN Number UN3089
Grade Reacton
Odor Odorless
Chemical Name or Material Samarium cobalt, Intermetallic, Magnet Alloy Additive
Hazard Category H228-H317-H334-H350
Hazard Statement GHS H Statement
H334-H228-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Flammable solid.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P210-P240-P241-P261-P272-P280g-P281-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: METAL POWDERS, FLAMMABLE, N.O.S.
EINECSNumber 234-625-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only