accessibility menu, dialog, popup

UserName

CAS RN 12017-68-4

IUPAC Name: pentacobalt samarium
Synonyms: pentacobalt samarium
Molecular Weight (g/mol): 445.03
Molecular Formula: Co5Sm
InChi Key: NDYCBWZIOSTTHS-UHFFFAOYSA-N
SMILES: [Co].[Co].[Co].[Co].[Co].[Sm]
Grade 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results