missing translation for 'onlineSavingsMsg'
Learn More

Salvianolic acid B, ≥95.0% (HPLC), MilliporeSigma™ Supelco™

Catalog No. 111012772
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111012772 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111012772 Supplier Merck Emd Millipore Supplier No. 4972410MG
Only null left

Salvianolic acid B is a cell protective antioxidant and free radical scavenger being investigated for a variety of conditions from ischemic heart disorders to Alzheimer's disease.

Food and Beverages

Specifications

Packaging Bottomless Glass Bottle with Inserted Fused Cone
CAS 121521-90-2
Physical Form Neat
Linear Formula C36H30O16
MDL Number MFCD23115783
Synonym (2 S,3 S)-4-[(1 E)-3-[(1 R)-1-Carboxy-2-(3,4-dihydroxyphenyl)ethoxy]-3-oxo-1-propen-1-yl]-2-(3,4-dihydroxyphenyl)-2,3-dihydro-7-hydroxy-3-benzofurancarboxylic acid 3-[(1 R)-1-carboxy-2-(3,4-dihydroxyphenyl)ethyl] ester; Dan Shen Suan B; Danfensuan B; Lithospermic acid B
Type Analytical Standard
Quantity 10 mg
Formula Weight 718.61
Percent Purity ≥95.0% (HPLC)
Grade Analytical Standard
Show More Show Less