missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eagle™ Salvage Poly Drum

Salvage drums are designed for use with damaged or leaking containers of hazardous materials, or for used absorbent materials contaminated with hazardous substances.
Specifications
| Product Type | Salvage Drum |
|---|---|
| Closure Included | Yes |
| Color | Yellow |
| For Use With (Application) | Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |
| Material | HDPE |
Description
- For use with damaged or leaking containers of hazardous materials or used absorbent materials contaminated with hazardous substances
- Salvage Drum labeling
- Passed a 3-PSI pressure test
Specifications
| Salvage Drum | |
| Yellow | |
| HDPE |
| Yes | |
| Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration |