missing translation for 'onlineSavingsMsg'
Learn More

Eagle™ Salvage Poly Drum
GREENER_CHOICE

Catalog No. 19338486
Change view
Click to view available options
Closure Type:
Metal Bolt Ring
Metal Lever-lock
Capacity (English):
20 gal.
65 gal.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Capacity (English)
19338486 Metal Bolt Ring 65 gal.
19338441 Metal Lever-lock 20 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19338486 Supplier Eagle™ Supplier No. 1665

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Salvage drums are designed for use with damaged or leaking containers of hazardous materials, or for used absorbent materials contaminated with hazardous substances.

  • For use with damaged or leaking containers of hazardous materials or used absorbent materials contaminated with hazardous substances
  • Salvage Drum labeling
  • Passed a 3-PSI pressure test

Specifications

Product Type Salvage Drum
Closure Included Yes
Closure Type Metal Bolt Ring
Capacity (English) 65 gal.
Capacity (Metric) 246.35 L
Color Yellow
For Use With (Application) Safe storage, handling and transportation of hazardous materials, spill containment and cleanup of hazardous materials, original shipping container, bio waste disposal and incineration
Material HDPE
Height (English) 38.88 in.
Height (Metric) 98.8 cm
Weight (English) 53 lb.
Show More Show Less