missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Salmeterol xinafoate

Catalog No. AC462171000
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC462171000 100 mg
AC462175000 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC462171000 Supplier Thermo Scientific Chemicals Supplier No. 462171000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 94749-08-3
Molecular Formula C36H45NO7
Molecular Weight (g/mol) 603.76
MDL Number MFCD00897708
InChI Key XTZNCVSCVHTPAI-UHFFFAOYNA-N
Synonym salmeterol xinafoate, salmetedur, arial, serevent diskus, asmerole, betamican, dilamax, inaspir, ultrabeta, beglan
PubChem CID 56801
IUPAC Name 2-(hydroxymethyl)-4-[1-hydroxy-2-[6-(4-phenylbutoxy)hexylamino]ethyl]phenol;1-hydroxynaphthalene-2-carboxylic acid
SMILES OC(=O)C1=CC=C2C=CC=CC2=C1O.OCC1=CC(=CC=C1O)C(O)CNCCCCCCOCCCCC1=CC=CC=C1

Specifications

Melting Point 126.0°C to 128.0°C
Quantity 100 mg
Solubility Information Solubility in water: soluble. Other solubilities: soluble in dmso, methanol
Formula Weight 603.75
Chemical Name or Material Salmeterol xinafoate

RUO – Research Use Only