missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ salm Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144457
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144457 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 missing translation for 'options'
Catalog No. FEPA5144457 missing translation for 'mfr' Invitrogen™ missing translation for 'supplierNo' PA5144457
Only null left

Rabbit Polyclonal Antibody

salm Polyclonal Antibody for Western Blot, ELISA

Required for the establishment of the posterior-most head and the anterior-most tail segments of the embryo. Probably function as a transcriptional regulator. Could repress the transcription of the tsh gene.

Specifications

Antigen salm
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% Proclin 300; pH 7.4
Gene salm
Gene Accession No. P39770
Gene Alias sal
Gene Symbols salm
Host Species Rabbit
Immunogen Recombinant Drosophila melanogaster (Fruit fly) SALM protein.
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 34569
Target Species Drosophila
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less