missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ SALL3 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA553467
Description
Immunogen sequence: AYDDKNAETL SSYDDDMDEN SMEDDAELKD AATDPAKPLL SYAGSCPPSP PSVISSIAAL ENQMKMIDSV MSCQQLTGLK SVENGSGE Highest antigen sequence identity to the following orthologs: Mouse - 83%, Rat - 83%.
SALL3 belongs to the sal C2H2-type zinc-finger protein family. It contains 9 C2H2-type zinc fingers. SALL3 is a probable transcription factor.Specifications
| SALL3 | |
| Polyclonal | |
| Unconjugated | |
| SALL3 | |
| B130022O04Rik; C2H2 zinc finger protein SALL3; hSALL3; MSal; Msal-1; Sal; Sall3; sal-like 3; sal-like 3 (Drosophila); sal-like protein 3; Salt; Spalt; spalt like transcription factor 3; spalt-like protein 3; spalt-like transcription factor 3; spalt-like zinc finger protein; zinc finger protein 796; Zinc finger protein SALL3; ZNF796 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 27164 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXA9 | |
| SALL3 | |
| Recombinant protein corresponding to Human SALL3. Recombinant protein control fragment (Product #RP-91647). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |