Learn More
Chemical Identifiers
| CAS | 552-94-3 |
|---|---|
| Molecular Formula | C14H10O5 |
| Molecular Weight (g/mol) | 258.22 |
| MDL Number | MFCD00020252 |
| InChI Key | WVYADZUPLLSGPU-UHFFFAOYSA-N |
| Synonym | salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal |
| PubChem CID | 5161 |
| ChEBI | CHEBI:9014 |
| IUPAC Name | 2-(2-hydroxybenzoyl)oxybenzoic acid |
| SMILES | C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity & Availability | ||||
|
AC133400250
|
N/A | 25g | Glass bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| C14H10O5 | |
| MFCD00020252 | |
| salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
| CHEBI:9014 | |
| C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O |
Specifications
| 552-94-3 | |
| 97.5% min. (HPLC) | |
| MFCD00020252 | |
| 15,8477 | |
| Solubility in water: hydrolysis. Other solubilities: soluble in alcohol and ether, sparingly soluble in benzene, slightly soluble in acetone | |
| C1=CC=C(C(=C1)C(=O)OC2=CC=CC=C2C(=O)O)O | |
| 258.22 | |
| CHEBI:9014 | |
| 98% |
| Authentic | |
| C14H10O5 | |
| 10,II,54 | |
| salsalate, sasapyrine, salicylsalicylic acid, disalicylic acid, disalcid, sasapyrinum, saloxium, 2-carboxyphenyl salicylate, salina, diacesal | |
| WVYADZUPLLSGPU-UHFFFAOYSA-N | |
| 2-(2-hydroxybenzoyl)oxybenzoic acid | |
| 5161 | |
| 258.22 | |
| Salicylsalicylic acid, 99% |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
EINECSNumber : 209-027-4
RTECSNumber : DH2080000
TSCA : TSCA
RUO – Research Use Only