missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Salicylic Acid, Crystal, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
$1,225.47 - $1,225.47
Chemical Identifiers
| CAS | 69-72-7 |
|---|---|
| Molecular Formula | C7H6O3 |
| Molecular Weight (g/mol) | 138.12 |
| MDL Number | MFCD00002439 |
| InChI Key | YGSDEFSMJLZEOE-UHFFFAOYSA-N |
| Synonym | salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone |
| PubChem CID | 338 |
| ChEBI | CHEBI:16914 |
| IUPAC Name | 2-hydroxybenzoic acid |
| SMILES | OC(=O)C1=CC=CC=C1O |
Chemical Identifiers
| 69-72-7 | |
| 138.12 | |
| YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
| 338 | |
| 2-hydroxybenzoic acid |
| C7H6O3 | |
| MFCD00002439 | |
| salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
| CHEBI:16914 | |
| OC(=O)C1=CC=CC=C1O |
Specifications
| 69-72-7 | |
| MFCD00002439 | |
| YGSDEFSMJLZEOE-UHFFFAOYSA-N | |
| 2-hydroxybenzoic acid | |
| 338 | |
| 138.12 | |
| 500 g | |
| Wide Mouth Amber Glass Bottle | |
| 1L = 1.44kg |
| C7H6O3 | |
| salicylic acid, o-hydroxybenzoic acid, 2-carboxyphenol, o-carboxyphenol, rutranex, salonil, retarder w, keralyt, duoplant, freezone | |
| OC(=O)C1=CC=CC=C1O | |
| 138.12 | |
| CHEBI:16914 | |
| BAKER ANALYZED™ ACS Reagent | |
| 157.00°C | |
| Crystalline | |
| Salicylic Acid |