missing translation for 'onlineSavingsMsg'
Learn More

Salicylaldehyde, Analytical Standard, ≥98.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111020593
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111020593 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111020593 Supplier Merck Emd Millipore Supplier No. 032731MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Specifications

CAS 90-02-8
Vapor Pressure 1 mmHg (33°C)
Vapor Density 4.2 (vs. air)
Density 1.146 g/mL (at 25°C)
Linear Formula 2-(HO)C6H4CHO
MDL Number MFCD00003317
Melting Point 1°C to 2°C
Boiling Point 197°C
Synonym 2-Hydroxybenzaldehyde
Quantity 1 mL
Formula Weight 122.12
Percent Purity ≥98.5% (GC)
Grade Analytical Standard
Show More Show Less