missing translation for 'onlineSavingsMsg'
Learn More

Salicyl Hydrazide 98.0+%, TCI America™

$200.21 - $200.21

Chemical Identifiers

CAS 936-02-7
Molecular Formula C7H8N2O2
Molecular Weight (g/mol) 152.153
MDL Number MFCD00007599
InChI Key XSXYESVZDBAKKT-UHFFFAOYSA-N
Synonym salicylhydrazide, 2-hydroxybenzhydrazide, salicyl hydrazide, salicyloyl hydrazide, salicylic acid hydrazide, salicylic hydrazide, salicyloylhydrazine, salicylohydrazide, o-hydroxybenzhydrazide, o-hydroxybenzoylhydrazide
PubChem CID 13637
IUPAC Name 2-hydroxybenzohydrazide
SMILES C1=CC=C(C(=C1)C(=O)NN)O
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S024225G
View Documents
TCI America
S024225G
25 g
Each for $200.21
Only null left
 
Description

Description

Specifications

Chemical Identifiers

936-02-7
152.153
XSXYESVZDBAKKT-UHFFFAOYSA-N
13637
C1=CC=C(C(=C1)C(=O)NN)O
C7H8N2O2
MFCD00007599
salicylhydrazide, 2-hydroxybenzhydrazide, salicyl hydrazide, salicyloyl hydrazide, salicylic acid hydrazide, salicylic hydrazide, salicyloylhydrazine, salicylohydrazide, o-hydroxybenzhydrazide, o-hydroxybenzoylhydrazide
2-hydroxybenzohydrazide

Specifications

936-02-7
White
MFCD00007599
salicylhydrazide, 2-hydroxybenzhydrazide, salicyl hydrazide, salicyloyl hydrazide, salicylic acid hydrazide, salicylic hydrazide, salicyloylhydrazine, salicylohydrazide, o-hydroxybenzhydrazide, o-hydroxybenzoylhydrazide
C1=CC=C(C(=C1)C(=O)NN)O
152.153
152.15
Crystalline Powder
150°C
C7H8N2O2
25 g
XSXYESVZDBAKKT-UHFFFAOYSA-N
2-hydroxybenzohydrazide
13637
≥98.0% (HPLC,T)
Salicyl Hydrazide
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-1665

RTECSNumber : VO3700000

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'