missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
| Salicin | |
| 200°C | |
| C13H18O7 | |
| 25 g | |
| NGFMICBWJRZIBI-UJPOAAIJSA-N | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol | |
| 439503 | |
| 286.28 | |
| Crystalline Powder |
| 138-52-3 | |
| White | |
| MFCD00006590 | |
| salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
| C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O | |
| 286.28 | |
| CHEBI:17814 | |
| ≥98.0% (HPLC) |
Chemical Identifiers
| 138-52-3 | |
| 286.28 | |
| NGFMICBWJRZIBI-UJPOAAIJSA-N | |
| 439503 | |
| (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol |
| C13H18O7 | |
| MFCD00006590 | |
| salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside | |
| CHEBI:17814 | |
| C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O |
Safety and Handling
RTECSNumber : LZ5901700
TSCA : Yes