missing translation for 'onlineSavingsMsg'
Learn More

Salicin 98.0+%, TCI America™

$65.46 - $188.67

Chemical Identifiers

CAS 138-52-3
Molecular Formula C13H18O7
Molecular Weight (g/mol) 286.28
MDL Number MFCD00006590
InChI Key NGFMICBWJRZIBI-UJPOAAIJSA-N
Synonym salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
PubChem CID 439503
ChEBI CHEBI:17814
IUPAC Name (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
SMILES C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
S00035G
View Documents
TCI America
S00035G
5 g
Each for $65.46
Only null left
 
S000325G
View Documents
TCI America
S000325G
25 g
Each for $188.67
Only null left
 
Description

Description

Specifications

Chemical Identifiers

138-52-3
286.28
NGFMICBWJRZIBI-UJPOAAIJSA-N
439503
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
C13H18O7
MFCD00006590
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
CHEBI:17814
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O

Specifications

138-52-3
White
MFCD00006590
salicin, salicoside, salicine, salicyl alcohol glucoside, d---salicin, 2r,3s,4s,5r,6s-2-hydroxymethyl-6-2-hydroxymethyl phenoxy tetrahydro-2h-pyran-3,4,5-triol, saligenin beta-d-glucopyranoside, 2-hydroxymethyl phenyl beta-d-glucopyranoside, d-salicin, saligenin-beta-d-glucopyranoside
C1=CC=C(C(=C1)CO)OC2C(C(C(C(O2)CO)O)O)O
286.28
CHEBI:17814
≥98.0% (HPLC)
Salicin
200°C
C13H18O7
5 g
NGFMICBWJRZIBI-UJPOAAIJSA-N
(2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-[2-(hydroxymethyl)phenoxy]oxane-3,4,5-triol
439503
286.28
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : LZ5901700

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'