missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ SalI (10 U/μL)

The SalI restriction enzyme recognizes G^TCGAC sites and cuts best at 37°C in O buffer.

$101.22 - $387.88

Specifications

Concentration 10 U/μL
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
FERER0642
View Documents
Thermo Scientific™
ER0642
5 x 1,500 units
Each for $387.88
Only null left
 
FERER0645
View Documents
Thermo Scientific™
ER0645
2,000 units
Each for $127.12
Only null left
 
FERER0641
View Documents
Thermo Scientific™
ER0641
1,500 units
Each for $101.22
Only null left
 
Description

Description

Lambda DNA digested with SalI, 0.7% agarose, 2 cleavage sites

conventional restriction endonucleases are a large collection of high quality restriction enzymes, optimized to work in one of the buffers of the Five Buffer System. In addition, the universal Tango buffer is provided for convenience in double digestions. All of the enzymes exhibit 100% activity in the recommended buffer and reaction conditions. To ensure consistent performance, Thermo Scientific restriction enzyme reaction buffers contain premixed BSA, which enhances the stability of many enzymes and binds contaminants that may be present in DNA preparations.

Highlights

  • Superior quality -stringent quality control and industry leading manufacturing process
  • Convenient color-coded Five Buffer System
  • Includes universal Tango buffer for double-digestions
  • BSA premixed in reaction buffers
  • Wide selection of restriction endonuclease specificities
Specifications

Specifications

10 U/μL
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.