missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol impurity I Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111014654
Change view
Click to view available options
Quantity:
20 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111014654 20 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111014654 Supplier Merck Emd Millipore Supplier No. PHR196220MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Salbutamol impurity I is a pharmaceutical impurity of salbutamol, belonging to the class of β2-agonists, widely used for the treatment of acute asthma symptoms and exercise-induced asthma in adults and children.

Specifications

CAS 56796-66-8
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H27 NO3
Synonym (1 RS)-2-[(1,1-Dimethylethyl)amino]-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol; 2-tert-Butylamino-1-[3-(hydroxymethyl)-4-benzyloxyphenyl]ethanol
Type Certified Reference Material
Quantity 20 mg
Formula Weight 329.43
Grade Certified Reference Material