missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol Impurity D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013245
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013245 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013245 Supplier Merck Emd Millipore Supplier No. PHR196050MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Pharmaceutical secondary standards for application in quality control provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards

Specifications

CAS 156339-88-7 (free base)
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula (C13H19 NO3)2 · H2 SO4
Synonym 5-[2-(tert-Butylamino)-1-hydroxyethyl]-2-hydroxybenzaldehyde hemisulfate salt; 5-{2-[(1,1-Dimethylethyl)amino]-1-hydroxyethyl}-2-hydroxybenzaldehyde hemisulfate salt
Type Certified Reference Material
Quantity 50 mg
Formula Weight 572.67
Grade Certified Reference Material