missing translation for 'onlineSavingsMsg'
Learn More

Salbutamol Impurity B, British Pharmacopoeia (BP) Reference Standard, MilliporeSigma™ Supelco™

Catalog No. 111022946
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111022946 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111022946 Supplier Merck Emd Millipore Supplier No. BP852

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the Authority of the issuing Pharmacopoeia.

Specifications

CAS 96948-64-0
Molecular Formula C12H19NO2
Linear Formula C12H19NO2
Synonym (1 RS)-2-[(1,1-Dimethylethyl)amino]-1-(4-hydroxyphenyl)ethanol; 2-tert-Butylamino-1-(4-hydroxyphenyl)ethanol
Type Pharmacopeia and Metrological Institutes Standards
Quantity 10 mg
Formula Weight 209.28
Grade British Pharmacopoeia (BP) Reference Standard