missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

Catalog No. S30046
Encompass
Change view
Click to view available options
Quantity:
1 Kit
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S30046 1 Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S30046 Supplier Invitrogen™ Supplier No. S30046
Season of Savings
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

SAIVI™ Rapid Antibody Labeling Kit, Alexa Fluor™ 750

The Alexa Fluor™ 750 SAIVI™ Antibody Labeling Kit provides a convenient means to label antibodies with an optimal degree of labeling for in vivo imaging applications (DOL; ∼2) over a 6-fold antibody concentration range with no adjustments in reaction volume, dye concentration, or antibody concentration necessary. Using this procedure, optimally labeled antibodies are ready for applications that require azide-free reagents, such as live-cell imaging or direct injection into animals.

Specifications

Product Type Labeling Kit
Quantity 1 Kit
Content And Storage Store in refrigerator 2°C to 8°C and protect from light.
Detection Method Fluorescence
Excitation/Emission 749/775
Labeling Scale 0.5 to 3 mg
Shipping Condition Room Temperature
Product Line Alexa Fluor, SAIVI
Label or Dye Alexa Fluor 750
Color Infrared
Labeling Target Antibodies
Label Type Alexa Fluor Dyes
Show More Show Less
What amount of conjugated antibody should I inject to image tumors?

A recommended starting dosage is 50 µg. You will need to determine the optimal dosage for your experimental model.

What size needle should I use for small animal in vivo imaging?

We recommend the use of a 28-32 gauge tuberculin or insulin syringe (0.3 or 1.0 mL volume) with a fixed (non-removable) needle.

What are the optimal and maximal volumes of reagent that can be injected into a mouse?

The volume of reagent that can be injected varies according to the route of administration. The following numbers are general guidelines for a 25 gram animal: Intravenous (IV)- 50-125 µl (recommended)- 200µl (maximum) ; Intraperitoneal (IP) 500µl (recommended) -2ml (maximum) ; Subcutaneous(SC) 100-250 µl (recommended)- 1ml (maximum).

What type of mice are the best for in vivo imaging?

Due to light scattering caused by fur, hairless mice such as athymic nude (nu/nu) mice are recommended for in vivo imaging. If this is not an option, the hair covering the area to be imaged should be removed using clippers or a chemical depilatory such as Nair depilatory.

For Research Use Only. Not for use in diagnostic procedures.