Learn More
Saint-Gobain Tygon S3™ E-3603 Flexible Tubings, Unipacs
Formulated for resistance to flex-fatigue and abrasion
$104.63 - $104.63
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity | |||
14-171-275
|
Saint-Gobain
ACF06017 |
1/4 in. | 3/8 in. | 1/16 in. |
Pack of 10 Each for $104.63
Case of 10 Pack for $1,046.30
|
|
|||
Description
- Bio-based formulation and contain no BPA or phthalates
- Provide high chemical resistance
- Non-oxidizing and non-contaminating
- Smooth, polished inner wall helps prevent buildup to facilitate cleaning
- Coils are marked at 30.4cm (1 ft.) intervals for easy measuring
- Slips easily over fittings and grips securely for simple lab set-ups
- Extra-heavy walls withstand full vacuum at room temperature (29.9 in. [759mm] of mercury at 23 °C [73 °F] and up to 27 in. [686mm] of mercury at 60°C [140°F])
- Sterilized by conventional autoclave methods (steam 30 min. at 15psi, 121°C) and Ethylene Oxide
- Instrumentation connection, vent, drain and other general laboratory applications
- Food and beverage
- Biopharmaceutical
- Analytical instruments
- Peristaltic and vacuum pumps
- Condensers
- Incubators
- Desiccators
- Gas lines
- Drain lines
Compliance
FDA, NSF, 3-A, meets USP Class VI, Japan Food Sanitation Law no. 370/1959, REACH, 1935/2004/EC and 10/2011/EU.
Specifications
| Clear |