missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Saint-Gobain Versilon™ Plasticizer-Free Tubing, Formulation 2001
Ideal for use in peristaltic pump systems, soap and detergent dispensing, ink transfer, water purification lines, chemical transfer, and food and beverage applications
$400.25
Specifications
| Certifications/Compliance | Compliant with FDA 21 CFR, 177.2600 |
|---|---|
| Overall Length (English) | 50 ft. |
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Diameter (English) Inner | Diameter (English) Outer | Thickness (English) | Price | Quantity & Availability | |||
14170103
|
Saint-Gobain
AE300017 |
1/4 in. | 3/8 in. | 1/16 in. |
Pack for $400.25
|
|
|||
Description
Versilon™ Plasticizer-Free Tubing, Formulation 2001 is uniquely engineered to provide flexibility, chemical resistance and extended pump life in a clear, plasticizer-/oil-free tubing product.
- Does not contaminate fluids
- Superior flex life in peristaltic pumps
- Chemically resistant to a wide range of fluids
- Crystal clear for easy visual flow monitoring
- Meets FDA criteria for food contact
- Will not degrade over time
- Will not release hazardous byproducts when properly incinerated
- Does not yield any taste
- Helps ensure accurate results from analytical instrument tests
†At 73°F (23°C).
Order Info
All products sold in 50-foot (15.24m) lengths
Compliance
Compliant with FDA 21 CFR, 177.2600
Specifications
| Compliant with FDA 21 CFR, 177.2600 | |
| 50 ft. |